Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2,2-difluoroacetamide, 98%

Catalog No. AAL0078514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0078514 25 g
AAL0078506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0078514 Supplier Thermo Scientific Chemicals Supplier No. L0078514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 354-28-9 | C2H2ClF2NO | 129.491 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354-28-9
Molecular Formula C2H2ClF2NO
Molecular Weight (g/mol) 129.491
MDL Number MFCD00039775
InChI Key PSAKKOKLSDIKEK-UHFFFAOYSA-N
Synonym chlorodifluoroacetamide, acetamide, 2-chloro-2,2-difluoro, difluorochloroacetamide, snylgqdjptadtaetp@, acmc-20algv, acetamide,2-difluoro, pubchem14583, 2-chloranyl-2,2-bis fluoranyl ethanamide
PubChem CID 67715
IUPAC Name 2-chloro-2,2-difluoroacetamide
SMILES C(=O)(C(F)(F)Cl)N

Specifications

Melting Point 79°C to 84°C
Quantity 25 g
Beilstein 1753808
Formula Weight 129.49
Percent Purity 98%
Chemical Name or Material 2-Chloro-2,2-difluoroacetamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-555-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.