Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-2,2-Difluoroacetophenone

Catalog No. 79237310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-237-310G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 79-237-310G Supplier Sigma Aldrich Supplier No. 79237310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloro-2,2-difluoroacetophenone is a difluorocarbene reagent, which is generally used in the synthesis of 2,2-difluoro enol silyl ethers, and gem-difluoromethene derived compounds.Reagent is an effective product for the synthesis of difluoromethylated phenols in the presence of mild base and aqueous solvent.

Specifications

Molecular Formula C22H22NO2PS
Refractive Index n20/D 1.4954; n20/D 1.497
Linear Formula C22H22NO2PS
Quantity 100 mg
Synonym 2-Chloro-2,2-difluoro-1-phenylethanone; Chlorodifluoromethyl phenyl ketone; alpha-Chloro-alpha,alpha-difluoroacetophenone
Molecular Weight (g/mol) 395.45 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.