Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2,5-dimethylhexane 98.0+%, TCI America™
SDP

Catalog No. C153310ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C153310ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C153310ML Supplier TCI America Supplier No. C153310ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29342-44-7
Molecular Formula C8H17Cl
Molecular Weight (g/mol) 148.674
MDL Number MFCD00060775
InChI Key SVRDRJZPEYTXKX-UHFFFAOYSA-N
PubChem CID 141477
IUPAC Name 2-chloro-2,5-dimethylhexane
SMILES CC(C)CCC(C)(C)Cl

Specifications

Color Colorless
Boiling Point 98°C
Quantity 10 mL
UN Number 1993
Formula Weight 148.67
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-2,5-dimethylhexane
EINECSNumber (2)-0066
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.