Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2',6'-dimethylacetanilide 98.0+%, TCI America™
SDP

Catalog No. C230725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C230725G 25 g
C23075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C230725G Supplier TCI America Supplier No. C230725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1131-01-7
Molecular Formula C10H12ClNO
Molecular Weight (g/mol) 197.662
MDL Number MFCD00000926
InChI Key FPQQSNUTBWFFLB-UHFFFAOYSA-N
Synonym 2-chloro-n-2,6-dimethylphenyl acetamide, 2-chloro-2',6'-dimethylacetanilide, n-chloroacetyl-2,6-dimethylaniline, 2-chloro-2',6'-acetoxylidide, chloroaceto-2,6-xylidide, acetamide, 2-chloro-n-2,6-dimethylphenyl, chloroacetamido-2,6-xylidine, 1-chloroacetylamino-2,6-dimethylbenzene, n-2,6-dimethylphenyl-2-chloroacetamide, unii-5d2b69oapx
PubChem CID 70798
IUPAC Name 2-chloro-N-(2,6-dimethylphenyl)acetamide
SMILES CC1=C(C(=CC=C1)C)NC(=O)CCl

Specifications

Melting Point 148°C
Quantity 25 g
Formula Weight 197.66
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-2′,6′-dimethylacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.