Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-chloro-2'-deoxyadenosine | 2627-62-5 | 98.5% | 285.69 g/mol | C10H12ClN5O3 | 10 MG
SDP

Catalog No. 5000286335
Encompass_Preferred

2'-Chloro-2'-deoxyadenosine is a purine nucleoside analogue used in laboratory research to study antitumor activity and DNA synthesis inhibition. It is supplied as a white to off-white solid in small milligram pack sizes for research use only. Follow the material safety data sheet for handling, storage, and disposal instructions.

  • Purity 98.5%.
  • Molecular weight 285.69 g/mol.
  • Chemical formula C10H12ClN5O3.
  • Appearance white to off-white solid.
  • Available in small milligram pack sizes (e.g., 5-200 mg).
  • Recommended storage: powder -20 °C (up to 3 years) or 4 °C; in solvent -80 °C (up to 6 months).

Catalog No. 50-002-86335 Supplier Medchemexpress LLC Supplier No. HY15233410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2'-Chloro-2'-deoxyadenosine is a purine nucleoside analogue used in laboratory research to study antitumor activity and DNA synthesis inhibition. It is supplied as a white to off-white solid in small milligram pack sizes for research use only. Follow the material safety data sheet for handling, storage, and disposal instructions.

  • Purity 98.5%.
  • Molecular weight 285.69 g/mol.
  • Chemical formula C10H12ClN5O3.
  • Appearance white to off-white solid.
  • Available in small milligram pack sizes (e.g., 5-200 mg).
  • Recommended storage: powder -20 °C (up to 3 years) or 4 °C; in solvent -80 °C (up to 6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.