Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2-methylbutane, 95%

Catalog No. aaa1170306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1170306 5 g
AAA1170314 25 g
AAA1170322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1170306 Supplier Thermo Scientific Chemicals Supplier No. A1170306
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 594-36-5 | C5H11Cl | 106.593 g/mol

2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents. Avoid extremes of temperature and direct sunlight.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-36-5
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00000818
InChI Key CRNIHJHMEQZAAS-UHFFFAOYSA-N
Synonym tert-amyl chloride, butane, 2-chloro-2-methyl, 2-methyl-2-chlorobutane, tert-pentyl chloride, 1,1-dimethylpropyl chloride, tertiary pentyl chloride, t-pentyl chloride, acmc-209mdc, 2-chloro-2-methyl butane, 2-chloro-2-methyl-butane
PubChem CID 61143
IUPAC Name 2-chloro-2-methylbutane
SMILES CCC(C)(C)Cl

Specifications

Melting Point -73°C
Density 0.866
Boiling Point 85°C to 86°C
Flash Point −10°C (14°F)
Refractive Index 1.405
Quantity 5 g
UN Number UN1107
Beilstein 1696930
Solubility Information Soluble in ethanol,ethyl ether. Insoluble in water.
Formula Weight 106.6
Percent Purity 95%
Chemical Name or Material 2-Chloro-2-methylbutane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES
EINECSNumber 209-836-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.