Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-2-methylpropane

Catalog No. C5635225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C5635225ML 25 mL
C563522L 2 L
C56352500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C5635225ML Supplier Sigma Aldrich Supplier No. C5635225ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloro-2-methylpropane

Specifications

CAS 507-20-0
Melting Point -25°C (lit.)
Density 0.851 g/mL (at 25°C (literature))
Boiling Point 51°C to 52°C (lit.)
Molecular Formula C4H9Cl
Refractive Index n20/D 1.385 (literature)
Linear Formula (CH3)3 CCl
MDL Number MFCD00000816
Quantity 25 mL
Synonym tert-Butyl chloride
Molecular Weight (g/mol) 92.57
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.