Learn More
Medchemexpress LLC 2-Chloro-2-methylpropane-d9 | 918-20-7 | C4D9Cl | 1 G
SDP
Catalog No. 5000375097
Encompass_Preferred
2-Chloro-2-methylpropane-d9 is a deuterium labeled compound designed for research applications. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. This product has a molecular weight of 101.62, appears as a colorless to light yellow liquid, and has a purity of 97.83%.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes of hydrogen, carbon, and other elements
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-75097 Supplier Medchemexpress LLC Supplier No. HY150519S1G
2-Chloro-2-methylpropane-d9 is a deuterium labeled compound designed for research applications. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. This product has a molecular weight of 101.62, appears as a colorless to light yellow liquid, and has a purity of 97.83%.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes of hydrogen, carbon, and other elements
  • Potential to affect pharmacokinetic and metabolic profiles of drugs