Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chloro-2-methylpropane-d9 | 918-20-7 | C4D9Cl | 100 MG
SDP

Catalog No. 5000375269
Encompass_Preferred

2-Chloro-2-methylpropane-d9 is a deuterium-labeled form of 2-Chloro-2-methylpropane. This compound serves as a tracer and internal standard in various analytical methods, including NMR, GC-MS, or LC-MS. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs, making this compound valuable in drug development processes.

  • Purity of 97.83%
  • Molecular weight of 101.62
  • Appears as a liquid with a colorless to light yellow hue
  • Functions as a tracer for quantitative analysis
  • Useful as an internal standard for NMR, GC-MS, or LC-MS
  • For research use only
  • Long-term storage at -20°C (3 years) or 4°C (2 years) for pure form
  • Short-term storage at -80°C (6 months) or -20°C (1 month) when in solvent

Catalog No. 50-003-75269 Supplier Medchemexpress LLC Supplier No. HY150519S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloro-2-methylpropane-d9 is a deuterium-labeled form of 2-Chloro-2-methylpropane. This compound serves as a tracer and internal standard in various analytical methods, including NMR, GC-MS, or LC-MS. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs, making this compound valuable in drug development processes.

  • Purity of 97.83%
  • Molecular weight of 101.62
  • Appears as a liquid with a colorless to light yellow hue
  • Functions as a tracer for quantitative analysis
  • Useful as an internal standard for NMR, GC-MS, or LC-MS
  • For research use only
  • Long-term storage at -20°C (3 years) or 4°C (2 years) for pure form
  • Short-term storage at -80°C (6 months) or -20°C (1 month) when in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.