Learn More
Medchemexpress LLC 2-Chloro-2-methylpropane-d9 | 918-20-7 | C4D9Cl | 500 MG
SDP
Catalog No. 5000375146
Encompass_Preferred
2-Chloro-2-methylpropane-d9 is the deuterium labeled 2-Chloro-2-methylpropane, intended for research use only. It serves as a tracer and internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Deuteration, involving stable heavy isotopes, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-75146 Supplier Medchemexpress LLC Supplier No. HY150519S500MG
2-Chloro-2-methylpropane-d9 is the deuterium labeled 2-Chloro-2-methylpropane, intended for research use only. It serves as a tracer and internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Deuteration, involving stable heavy isotopes, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Has potential to affect pharmacokinetic and metabolic profiles of drugs