Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2-oxo-1,3,2-dioxaphospholane 95.0+%, TCI America™
SDP

Catalog No. C125025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C125025G 25 g
C12505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C125025G Supplier TCI America Supplier No. C125025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6609-64-9
Molecular Formula C2H4ClO3P
Molecular Weight (g/mol) 142.475
MDL Number MFCD00043138
InChI Key SBMUNILHNJLMBF-UHFFFAOYSA-N
Synonym 2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphate
PubChem CID 81087
IUPAC Name 2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide
SMILES C1COP(=O)(O1)Cl

Specifications

Color Yellow
Boiling Point 91°C
Quantity 25 g
UN Number 3265
Formula Weight 142.47
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-oxo-1,3,2-dioxaphospholane
EINECSNumber (9)-0448
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.