Learn More
Matrix Scientific 2-CHLORO-2-OXOETHYL ACETAT-25G
Catalog No. 50281585
Encompass_Preferred
2-Chloro-2-oxoethyl acetate, -25g,C4H5ClO3, MFCD00011535, mw 136.54,
Catalog No. 50-281-585 Supplier Matrix Scientific Supplier No. 06347325G
2-Chloro-2-oxoethyl acetate, -25g,C4H5ClO3, MFCD00011535, mw 136.54,