Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-2-propen-1-ol

Catalog No. 3639015G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-390-15G 5 g
36-390-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-390-15G Supplier Sigma Aldrich Supplier No. 3639015G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-chloro-2-propen-1-ol is reported to undergo photodissociation at 193nm to generate ch2cch2oh radical intermediate
  • 2-chloro-2-propen-1-ol is formed as major product during base mediated reaction of 1,2,3-trichloropropane
  • 2-chloro-2-propen-1-ol is reported to react with phosphorus trichloride to yield phosphorous esters, while with phosphory chloride it yields phosphoric ester.2-chloro-2-propen-1-ol (2-chloropropenol) may be employed as carbon supplement for the growth of pseudomonas strains
  • It may be used in the preparation of 2-(4-octylphenyl)prop-2-en-1-ol

Specifications

CAS 5976-47-6
Density 1.162 g/mL (at 25°C (literature))
Boiling Point 133°C to 134°C (lit.)
Molecular Formula C3H5ClO
Refractive Index n20/D 1.459 (literature)
Linear Formula H2C=C(Cl)CH2OH
MDL Number MFCD00004701
Quantity 5 g
Synonym 2-Chloroallyl alcohol
Molecular Weight (g/mol) 92.52
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.