Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-3-(1-pyrrolidinylcarbonyl)pyridine

Catalog No. VBP000451G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VBP000451G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. VBP000451G Supplier Sigma Aldrich Supplier No. VBP000451G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-nitroanisole undergoes photochemical nucleophilic aromatic substitution by hydroxide ion to form 4-methoxyphenol and 4-nitrophenol.4-nitroanisole was used as probe to determine +ß* of kamlet-taft solvent parameters and high pressure and supercritical water in the temperature range of 16-420 c
  • 4-nitroanisole was used as carbon and energy supplement for the isolation of rhodococcus strains.4-nitroanisole is o-demethylated to 4-nitrophenol by human liver microsomes

Specifications

Molecular Formula C10H11ClN2O
Linear Formula C10H11ClN2O
MDL Number MFCD01342311
Quantity 25 g
Molecular Weight (g/mol) 210.66
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.