Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3,5-bis(trifluoromethyl)aniline, 98%, Thermo Scientific™

Catalog No. AAH6411906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6411906 5 g
AAH6411914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6411906 Supplier Thermo Scientific Chemicals Supplier No. H6411906

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-3,5-bis(trifluoromethyl)aniline is used as dyestuff Intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201593-90-0
Molecular Formula C8H4ClF6N
Molecular Weight (g/mol) 263.567
MDL Number MFCD04038716
InChI Key JKUFETFGEJPHEM-UHFFFAOYSA-N
Synonym 2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine
PubChem CID 2757429
IUPAC Name 2-chloro-3,5-bis(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.595 g/mL
Refractive Index 1.459
Quantity 5 g
UN Number UN2810
Beilstein 13730810
Solubility Information Slightly soluble in water.
Formula Weight 263.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-3,5-bis(trifluoromethyl)aniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.