Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3,5-dinitrobenzoic acid, 97% (dry wt.), may cont. up to ca 5% water

Catalog No. AAA1567514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1567514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1567514 Supplier Thermo Scientific Chemicals Supplier No. A1567514

missing translation for 'validMainframeMsg'

CAS: 2497-91-8 | C7H3ClN2O6 | 246.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2497-91-8
Molecular Formula C7H3ClN2O6
Molecular Weight (g/mol) 246.56
MDL Number MFCD00007070
InChI Key ADTKEYLCJYYHHH-UHFFFAOYSA-N
Synonym 2-chloro-3,5-dinitrobenzoic acid, benzoic acid, 2-chloro-3,5-dinitro, 2-carboxy-4,6-dinitrochlorobenzene, 3,5-dinitro-2-chlorobenzoic acid, acmc-209uud, timtec-bb sbb003177, wln: wnr bg cvq enw, 4-09-00-01359 beilstein handbook reference, ksc495o8b, 2-chloro-3,5-dinitro-benzoicaci
PubChem CID 17247
IUPAC Name 2-chloro-3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=CC(=C1Cl)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 196°C to 201°C
Quantity 25 g
Beilstein 2146480
Formula Weight 246.56
Percent Purity 97%
Assay (dry wt.), may cont. up to ≈5% water
Chemical Name or Material 2-Chloro-3,5-dinitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-684-9
RTECSNumber DG4996500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.