Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-cyanopyridine 97.0+%, TCI America™
SDP

Catalog No. C136925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C136925G 25 g
C1369500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C136925G Supplier TCI America Supplier No. C136925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6602-54-6
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.554
MDL Number MFCD00014628
InChI Key JAUPUQRPBNDMDT-UHFFFAOYSA-N
Synonym 2-chloro-3-cyanopyridine, 2-chloronicotinonitrile, 2-chloro-3-pyridinecarbonitrile, 3-cyano-2-chloropyridine, 3-pyridinecarbonitrile, 2-chloro, 2-chloro-nicotinonitrile, 2-chloronicotinitrile, 2-chloro-3-cyano-pyridine, 2-chloro-3-cyanopyridine 2-chloronicotinonitrile, 2chloronicotinonitrile
PubChem CID 81079
IUPAC Name 2-chloropyridine-3-carbonitrile
SMILES C1=CC(=C(N=C1)Cl)C#N

Specifications

Melting Point 106°C
Color White
Quantity 25 g
UN Number 3439
Formula Weight 138.55
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-cyanopyridine
EINECSNumber (5)-6782
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.