Learn More
Cayman Chemical 2-chloro-3-DeazaadenosIn 10mg
SDP
Catalog No. 502894911
Encompass_Preferred
A stable analog of adenosine that acts as an agonist for adenosine receptors (Ki = 0.3, 0.08, 25.5, and 1.9 μM for A1, A2A, A2B, and A3 receptors, respectively); modulates neurotransmission release and blocks seizures by activating adenosine receptors
Catalog No. 50-289-4911 Supplier Cayman Chemical Supplier No. 900078710 MG
A stable analog of adenosine that acts as an agonist for adenosine receptors (Ki = 0.3, 0.08, 25.5, and 1.9 μM for A1, A2A, A2B, and A3 receptors, respectively); modulates neurotransmission release and blocks seizures by activating adenosine receptors