Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-fluoropyridine, 97%

Catalog No. AAL2002203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2002203 1 g
AAL2002206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2002203 Supplier Thermo Scientific Chemicals Supplier No. L2002203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 17282-04-1 | C5H3ClFN | 131.534 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17282-04-1
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD03095302
InChI Key SVAZIMBLBHOVIR-UHFFFAOYSA-N
Synonym 3-fluoro-2-chloropyridine, 2-chloro-3-fluoro pyridine, pyridine, 2-chloro-3-fluoro, 2-chloro-3-fluoro-pyridine, pubchem2532, acmc-1cc0d, ksc174s3h, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g
PubChem CID 2782791
IUPAC Name 2-chloro-3-fluoropyridine
SMILES C1=CC(=C(N=C1)Cl)F

Specifications

Density 1.325
Boiling Point 62°C to 64°C (23 mmHg)
Flash Point 72°C (161°F)
Odor Unpleasant
Refractive Index 1.5095
Quantity 1 g
UN Number UN2810
Beilstein 1363435
Formula Weight 131.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-fluoropyridine
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H310-H330-H318-H227-H315-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.