Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-iodopyridine, 97%, Thermo Scientific™

Catalog No. AAL2002806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2002806 5 g
AAL2002803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2002806 Supplier Thermo Scientific Chemicals Supplier No. L2002806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78607-36-0
Molecular Formula C5H3ClIN
Molecular Weight (g/mol) 239.44
MDL Number MFCD00661298
InChI Key OHWSWGXNZDSHLM-UHFFFAOYSA-N
Synonym 2-chloro-3-iodo-pyridine, pyridine, 2-chloro-3-iodo, 3-iodo-2-chloropyridine, zlchem 306, pubchem1188, acmc-209pes, 2-chloro-3-iodopyridine, ksc378k0b, 2-chloranyl-3-iodanyl-pyridine, 2-choro-3-iodopyridine
PubChem CID 2735743
IUPAC Name 2-chloro-3-iodopyridine
SMILES C1=CC(=C(N=C1)Cl)I

Specifications

Melting Point 93°C to 97°C
Quantity 5 g
Beilstein 4243304
Sensitivity Light sensitive
Formula Weight 239.44
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-iodopyridine
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.