Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-3-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
18368-76-8
C6H6ClN
127.57
MFCD00234177
RKVUCIFREKHYTL-UHFFFAOYSA-N
2-chloro-3-picoline,2-chloro 3-methylpyridine,3-picoline, 2-chloro,pyridine, 2-chloro-3-methyl,2-chloro-3-methyl-pyridine,pyridine,2-chloro-3-methyl,pubchem2074,2-chloro-methyl-pyridine,2-chloro3-methyl-pyridine
87603
2-chloro-3-methylpyridine
CC1=C(N=CC=C1)Cl
18368-76-8
C6H6ClN
127.57
MFCD00234177
RKVUCIFREKHYTL-UHFFFAOYSA-N
2-chloro-3-picoline,2-chloro 3-methylpyridine,3-picoline, 2-chloro,pyridine, 2-chloro-3-methyl,2-chloro-3-methyl-pyridine,pyridine,2-chloro-3-methyl,pubchem2074,2-chloro-methyl-pyridine,2-chloro3-methyl-pyridine
87603
2-chloro-3-methylpyridine
CC1=C(N=CC=C1)Cl
Flash Point 79°C
Specific Gravity 1.17
Refractive Index 1.5320 to 1.534
Boiling Point 192°C to 193°C
Quantity 5g
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.1700g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 127.57
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Chloro-3-picoline
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 242-242-1

Recommended Storage : this product may darken during storage