Learn More
Medchemexpress LLC 2-Chloro-3-pyridinol | 6636-78-8 | 99.9% | 129.55 | 100 G
SDP
Catalog No. 5000403772
Encompass_Preferred
2-Chloro-3-pyridinol is a biochemical reagent useful in life science research and for synthesizing active compounds. It is also found as a halopyridinol disinfection by-product in drinking water samples.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Can be used in the synthesis of other active compounds
  • Low cytotoxicity to human hepatocellular carcinoma HepG2 cells
Catalog No. 50-004-03772 Supplier Medchemexpress LLC Supplier No. HY32833100G
2-Chloro-3-pyridinol is a biochemical reagent useful in life science research and for synthesizing active compounds. It is also found as a halopyridinol disinfection by-product in drinking water samples.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Can be used in the synthesis of other active compounds
  • Low cytotoxicity to human hepatocellular carcinoma HepG2 cells