Learn More
Medchemexpress LLC 2-Chloro-3-pyridinol | 6636-78-8 | 99.9% | 129.55 | 25 G
SDP
Catalog No. 5000403331
Encompass_Preferred
2-Chloro-3-pyridinol is a biochemical reagent useful as a biological material or organic compound for life science-related research. It can also be used in the synthesis of other active compounds. This compound is also identified as a halopyridinol disinfection by-product in drinking water samples. In vitro studies show that 2-Chloro-3-pyridinol exhibits very low cytotoxicity to human hepatocellular carcinoma HepG2 cells, with an EC50 > 10000 μM.
  • Can be used as a biological material or organic compound for research
  • Useful in the synthesis of other active compounds
  • Exhibits very low cytotoxicity to human hepatocellular carcinoma HepG2 cells
Catalog No. 50-004-03331 Supplier Medchemexpress LLC Supplier No. 25GHY32833
2-Chloro-3-pyridinol is a biochemical reagent useful as a biological material or organic compound for life science-related research. It can also be used in the synthesis of other active compounds. This compound is also identified as a halopyridinol disinfection by-product in drinking water samples. In vitro studies show that 2-Chloro-3-pyridinol exhibits very low cytotoxicity to human hepatocellular carcinoma HepG2 cells, with an EC50 > 10000 μM.
  • Can be used as a biological material or organic compound for research
  • Useful in the synthesis of other active compounds
  • Exhibits very low cytotoxicity to human hepatocellular carcinoma HepG2 cells