Learn More
Medchemexpress LLC 2-Chloro-3-pyridinol | 6636-78-8 | 99.9% | 129.55 | 5 G
SDP
Supplier:  Medchemexpress LLC HY328335G
Encompass_Preferred
2-Chloro-3-pyridinol is a biochemical reagent suitable for use as a biological material or organic compound in life science research. It can also be utilized in the synthesis of other active compounds. Additionally, it is identified as a halopyridinol disinfection byproduct found in drinking water samples. For research use only, not for sale to patients.
  • Biochemical reagent
  • Used as a biological material
  • Used as an organic compound in life science research
  • Utilized in the synthesis of other active compounds
  • Identified as a halopyridinol disinfection byproduct
  • For research use only
Catalog No. 50-004-04038