Learn More
Medchemexpress LLC 2-Chloro-3-pyridinol | 6636-78-8 | 99.9% | 129.55 | 50 G
SDP
Supplier:  Medchemexpress LLC HY3283350G
Encompass_Preferred
2-Chloro-3-pyridinol is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It can also be used in the synthesis of other active compounds. In addition, 2-Chloro-3-pyridinol is a halopyridinol disinfection by-product in drinking water samples. For research use only. We do not sell to patients.
  • Exhibits very low cytotoxicity to human hepatocellular carcinoma HepG2 cells, with an EC50 > 10000 μM.
Catalog No. 50-004-01445