Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-(trifluoromethyl)benzylamine 95.0+%, TCI America™
SDP

Catalog No. C29351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29351G Supplier TCI America Supplier No. C29351G
Add to Cart
Add to Cart

Specifications

CAS 39226-96-5
Melting Point 38°C
Color Yellow
Boiling Point 223°C
Molecular Formula C8H7ClF3N
Quantity 1 g
MDL Number MFCD04115990
UN Number 3259
Synonym 2-chloro-3-trifluoromethyl phenyl methanamine, 2-chloro-3-trifluoromethyl benzylamine, 1-2-chloro-3-trifluoromethyl phenyl methanamine, 2-chloro-3-trifluoromethyl-benzylamine, 2-chloro-3-trifluoromethyl phenyl methyl amine, benzenemethanamine, 2-chloro-3-trifluoromethyl, pubchem23903, acmc-1afeh, ksc845q8b
InChI Key GRFVQNWTQVWNBY-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)C(F)(F)F)Cl)CN
IUPAC Name [2-chloro-3-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol) 209.596
PubChem CID 5224207
Formula Weight 209.60
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzylamine
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.