Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. C119325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C119325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C119325G Supplier TCI America Supplier No. C119325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65753-47-1
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.542
MDL Number MFCD00042223
InChI Key RXATZPCCMYMPME-UHFFFAOYSA-N
Synonym 2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t
PubChem CID 589833
IUPAC Name 2-chloro-3-(trifluoromethyl)pyridine
SMILES C1=CC(=C(N=C1)Cl)C(F)(F)F

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 76°C
Quantity 25 g
UN Number 2811
Formula Weight 181.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)pyridine
EINECSNumber (5)-2954
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.