Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-3-(trimethylsilyl)pyridine

Catalog No. SYX003161G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003161G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003161G Supplier Sigma Aldrich Supplier No. SYX003161G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-chloro-3-methylpentane(3c3mp) has structure similar to tertiary chloride groups which are formed as structural defects during the radical polymerization of vinyl chloride (vc)
  • Therefore, 3c3mp is an ideal model compound to study if tertiary chloride defect present in commercial pvc can be active initiating species for metal-catalyzed living radical graft copolymerization
  • 3c3mp can also be used as a reactant to prepare chloro(1-ethyl-1-methylpropyl)magnesium (grignard reagent), which is used in the kumada-tamao-corriu coupling reaction

Specifications

CAS 77332-76-4
Molecular Formula C8H12ClNSi
Linear Formula C8H12ClNSi
MDL Number MFCD09763704
Quantity 25 g
Molecular Weight (g/mol) 185.73
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.