Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4,6-dibromoaniline 99.0+%, TCI America™
SDP

Catalog No. C138510G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C138510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C138510G Supplier TCI America Supplier No. C138510G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 874-18-0
Molecular Formula C6H4Br2ClN
Molecular Weight (g/mol) 285.363
MDL Number MFCD00031419
InChI Key MFROLNRQDCAZCD-UHFFFAOYSA-N
Synonym 2-chloro-4,6-dibromoaniline, 6-chloro-2,4-dibromoaniline, 4,6-dibromo-2-chlorophenylamine, pubchem3631, acmc-209qkg, 4,6-dibromo-2-chloroaniline, benzenamine,2,4-dibromo-6-chloro, labotest-bb lt01147812, 2,4-dibromo-6-chloroaniline, 2,4-bis bromanyl-6-chloranyl-aniline
PubChem CID 4295982
IUPAC Name 2,4-dibromo-6-chloroaniline
SMILES C1=C(C=C(C(=C1Br)N)Cl)Br

Specifications

Melting Point 100°C
Quantity 10 g
Formula Weight 285.36
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,6-dibromoaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.