Learn More
Medchemexpress LLC 2-chloro-4,6-dimethoxy-1,3,5-triazine | 3140-73-6 | MFCD00075607 | 98.9% | 175.57 g/mol | C5H6ClN3O2 | 100 G
SDP
Catalog No. 5000301152
Encompass_Preferred
2-Chloro-4,6-dimethoxy-1,3,5-triazine is an s-triazine reagent widely used to activate carboxylic acids for amide and ester formation. It functions as a coupling reagent in peptide and organic synthesis and is employed as an intermediate for preparing activated triazinyl derivatives. Batch certificates and supplier listings report high assay values suitable for laboratory synthesis and method development.
  • Acts as an efficient coupling reagent for amide and ester formation.
  • Suitable for peptide synthesis and purification workflows.
  • High assay by qNMR (98.9%).
  • Enables preparation of activated triazinyl reagents and trialkylammonium salts.
  • Stable yet reactive under typical coupling conditions.
Catalog No. 50-003-01152 Supplier Medchemexpress LLC Supplier No. HYW016718100G
2-Chloro-4,6-dimethoxy-1,3,5-triazine is an s-triazine reagent widely used to activate carboxylic acids for amide and ester formation. It functions as a coupling reagent in peptide and organic synthesis and is employed as an intermediate for preparing activated triazinyl derivatives. Batch certificates and supplier listings report high assay values suitable for laboratory synthesis and method development.
  • Acts as an efficient coupling reagent for amide and ester formation.
  • Suitable for peptide synthesis and purification workflows.
  • High assay by qNMR (98.9%).
  • Enables preparation of activated triazinyl reagents and trialkylammonium salts.
  • Stable yet reactive under typical coupling conditions.