Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-4-aminotoluene

Catalog No. 23632250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-632-250G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-632-250G Supplier Sigma Aldrich Supplier No. 23632250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloro-4-aminotoluene

Specifications

CAS 95-74-9
Melting Point 24°C to 25°C (lit.)
Density 1.167 g/mL (at 25°C)
Boiling Point 237°C to 238°C (lit.)
Molecular Formula C7H8ClN
Refractive Index n20/D 1.584 (literature)
Linear Formula ClC6H3(CH3)NH2
MDL Number MFCD00007773
Quantity 250 mg
Synonym 3-Chloro-4-methylaniline; 3-Chloro-p-toluidine; 4-Amino-2-chlorotoluene
Molecular Weight (g/mol) 141.6
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.