Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-bromo-5-fluorobenzaldehyde, 98%

Catalog No. p-7055616
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6423803 1 g
AAH6423806 5 g
AAH64238MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6423803 Supplier Thermo Scientific Chemicals Supplier No. H6423803
Only null left
Add to Cart

CAS: 1214386-29-4 | C7H3BrClFO | 237.452 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214386-29-4
Molecular Formula C7H3BrClFO
Molecular Weight (g/mol) 237.452
MDL Number MFCD13185877
InChI Key XPEGZWLIWARWAA-UHFFFAOYSA-N
Synonym 2-chloro-4-bromo-5-fluorobenzaldehyde
PubChem CID 70680695
IUPAC Name 4-bromo-2-chloro-5-fluorobenzaldehyde
SMILES C1=C(C(=CC(=C1F)Br)Cl)C=O

Specifications

Color White
Quantity 1 g
Formula Weight 237.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-4-bromo-5-fluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.