Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Chloro-4-dimethylaminoazobenzene 98.0+%, TCI America™
SDP

Catalog No. C015725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C015725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C015725G Supplier TCI America Supplier No. C015725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3010-47-7
Molecular Formula C14H14ClN3
Molecular Weight (g/mol) 259.737
MDL Number MFCD00059478
InChI Key AXTUMSFTJSYZOD-UHFFFAOYSA-N
PubChem CID 18167
IUPAC Name 4-[(2-chlorophenyl)diazenyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2Cl

Specifications

Melting Point 111°C
Quantity 25 g
Formula Weight 259.74
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Chloro-4-dimethylaminoazobenzene
RTECSNumber BX5050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.