Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-fluorophenol, 98+%, Thermo Scientific™

Catalog No. AAA1086406
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1086406 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1086406 Supplier Thermo Scientific Chemicals Supplier No. A1086406

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1996-41-4
Density 1.345
Boiling Point 171°C to 172°C
Flash Point 75°C (167°F)
Odor Phenol-like
Molecular Formula C6H4ClFO
Refractive Index 1.53
Quantity 5g
MDL Number MFCD00002168
UN Number UN3265
Beilstein 1861277
Synonym phenol, 2-chloro-4-fluoro, 2-chlor-4-fluorophenol, 4-fluoro-2-chlorophenol, 2-chloro-4-fluoro-phenol, pubchem2372, 2-chloro-4-fluorophenol, acmc-209f4h, ksc493k3h, 1-bromo-2,3,5-trifluorobezene
InChI Key IGYXYGDEYHNFFT-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)Cl)O
IUPAC Name 2-chloro-4-fluorophenol
Molecular Weight (g/mol) 146.545
PubChem CID 74814
Formula Weight 146.55
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-4-fluorophenol
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 217-876-7
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.