Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-fluorophenol 98.0+%, TCI America™
SDP

Catalog No. C159825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C159825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C159825G Supplier TCI America Supplier No. C159825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1996-41-4
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.545
MDL Number MFCD00002168
InChI Key IGYXYGDEYHNFFT-UHFFFAOYSA-N
Synonym phenol, 2-chloro-4-fluoro, 2-chlor-4-fluorophenol, 4-fluoro-2-chlorophenol, 2-chloro-4-fluoro-phenol, pubchem2372, 2-chloro-4-fluorophenol, acmc-209f4h, ksc493k3h, 1-bromo-2,3,5-trifluorobezene
PubChem CID 74814
IUPAC Name 2-chloro-4-fluorophenol
SMILES C1=CC(=C(C=C1F)Cl)O

Specifications

Color White-Yellow
Boiling Point 172°C
Quantity 25 g
Formula Weight 146.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-4-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.