Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-fluoropyridine 98.0+%, TCI America™
SDP

Catalog No. C26275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26275G Supplier TCI America Supplier No. C26275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent used as a building block in pharmaceutical synthesis.

Chemical Identifiers

CAS 34941-91-8
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD04039345
InChI Key FGSAQRJRWCZLOB-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-4-fluoro, 2-chloro-4-fluoro pyridine, 2-chloro-4-fluoro-pyridine, pubchem6180, acmc-1aft7, 2-chlro-4-fluoroxypyridine, ksc222e0n, pyrimidine, 4-chloro-6-fluoro, abbypharma ap-14-5731
PubChem CID 2782792
IUPAC Name 2-chloro-4-fluoropyridine
SMILES C1=CN=C(C=C1F)Cl

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
UN Number 1993
Formula Weight 131.53
Percent Purity ≥98.0% (GC)
Grade Laboratory
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-fluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.