Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-iodo-3-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. C33681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33681G 1 g
C33685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33681G Supplier TCI America Supplier No. C33681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 153034-88-9
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.467
MDL Number MFCD03095293
InChI Key WHCRABJNJRAJIP-UHFFFAOYSA-N
Synonym 2-chloro-4-iodo-3-picoline, 2-chloro-3-methyl-4-iodopyridine, 2-chloro-4-iodo-3-methyl-pyridine, pyridine, 2-chloro-4-iodo-3-methyl, 2-chloro-4-iodo-3-methylpyridne, pubchem17059, 2-chloranyl-4-iodanyl-3-methyl-pyridine, 2-chloro-4-iodo-3-methylpyridine, 2-chloro-4-iodo-3-methyl-pyridine; 2-chloro-4-iodo-3-picoline
PubChem CID 10083628
IUPAC Name 2-chloro-4-iodo-3-methylpyridine
SMILES CC1=C(C=CN=C1Cl)I

Specifications

Melting Point 106°C
Quantity 1 g
Formula Weight 253.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-iodo-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.