Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-iodopyridine 98.0+%, TCI America™
SDP

Catalog No. C20895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20895G Supplier TCI America Supplier No. C20895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity building block for chemical synthesis with 98.0% GC purity.

Chemical Identifiers

CAS 153034-86-7
Molecular Formula C5H3ClIN
Molecular Weight (g/mol) 239.44
MDL Number MFCD01861983
InChI Key KJKIPRQNFDUULB-UHFFFAOYSA-N
Synonym 2-chloro-4-iodo-pyridine, 4-iodo-2-chloropyridine, 2-chloro-iodopyridine, pyridine, 2-chloro-4-iodo, zlchem 826, pubchem2579, acmc-209d8n, 2-chloro-4-iodo pyridine, ksc174s0f, 2-chloro-4-iodopyridine
PubChem CID 1516511
IUPAC Name 2-chloro-4-iodopyridine
SMILES C1=CN=C(C=C1I)Cl

Specifications

Melting Point 44°C
Color Yellow
Quantity 5 g
Formula Weight 239.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-iodopyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.