Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chloro-4-methoxypyridine | 17228-69-2 | 143.57 | 25 G
SDP

Catalog No. 5000449903
Encompass_Preferred

2-Chloro-4-methoxypyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • CAS number: 17228-69-2
  • Molecular formula: C6H6ClNO
  • Molecular weight: 143.57
  • Purity: 99.99% (GC)
  • Appearance: Colorless to off-white liquid
  • ¹H NMR spectrum: Consistent with structure
  • Storage (pure form): 3 years at -20°C, 2 years at 4°C
  • Storage (in solvent): 6 months at -80°C, 1 month at -20°C

Catalog No. 50-004-49903 Supplier Medchemexpress LLC Supplier No. 25GHYW007914
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloro-4-methoxypyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • CAS number: 17228-69-2
  • Molecular formula: C6H6ClNO
  • Molecular weight: 143.57
  • Purity: 99.99% (GC)
  • Appearance: Colorless to off-white liquid
  • ¹H NMR spectrum: Consistent with structure
  • Storage (pure form): 3 years at -20°C, 2 years at 4°C
  • Storage (in solvent): 6 months at -80°C, 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.