Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2-Chloro-4-methoxypyridine, 98%, [17228-69-2], Y15202 143.571 C6H6ClNO

Catalog No. 501456349
Encompass_Preferred

Compound: 2-Chloro-4-methoxypyridine

CAS#: 17228-69-2

MDL#: not listed

Background: A chlorinated, methoxylated pyridine used as a common building block in pharmaceutical research. The chlorine substituent supports nucleophilic aromatic substitution and cross-coupling chemistry.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-145-6349 Supplier Frontier Specialty Chemicals Supplier No. Y152021G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Chloro-4-methoxypyridine

CAS#: 17228-69-2

MDL#: not listed

Background: A chlorinated, methoxylated pyridine used as a common building block in pharmaceutical research. The chlorine substituent supports nucleophilic aromatic substitution and cross-coupling chemistry.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.