Learn More
AdipoGen 2-Chloro-4-methoxyresorcinol
SDP
Supplier:  AdipoGen CDXC0764G001
Encompass_Preferred
Chemical. CAS 81742-06-5. Formula C7H7ClO3. MW 174.58. Building block and intermediate used in synthesis of fluorescent probes, such as carboxy-fluoresceins.
Catalog No. 50-256-1339