Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-methyl-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. C20125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20125G 5 g
C20121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20125G Supplier TCI America Supplier No. C20125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23056-39-5 | C6H5ClN2O2 | 172.568 g/mol

Chemical Identifiers

CAS 23056-39-5
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
InChI Key JHARVUVBTAAPLA-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-4-picoline, pyridine, 2-chloro-4-methyl-3-nitro, 2-chloro-3-nitro-4-methylpyridine, 2-chloro-4-methyl-3-nitro-pyridine, pubchem6148, acmc-1cui4, 2-chloro-3-nitro4-picoline, ksc496c3n, 2-chlor-4-methyl-3-nitropyridin
PubChem CID 345363
IUPAC Name 2-chloro-4-methyl-3-nitropyridine
SMILES CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 53°C
Color Yellow
Quantity 5 g
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-methyl-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.