Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-methyl-5-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. C20211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20211G 1 g
C20215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20211G Supplier TCI America Supplier No. C20211G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23056-33-9
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD00010688
InChI Key HWZUMEVIIGNXGM-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-4-picoline, 2-chloro-4-methyl-5-nitro-pyridine, 2-chloro-4-methyl-5-nitro pyridine, 2-chloro-5-nitro-4-methylpyridine, pyridine, 2-chloro-4-methyl-5-nitro, pubchem6223, acmc-209g0j, ksc201s3j, tpc-py010
PubChem CID 345364
IUPAC Name 2-chloro-4-methyl-5-nitropyridine
SMILES CC1=CC(Cl)=NC=C1[N+]([O-])=O

Specifications

Melting Point 38°C
Boiling Point 91°C
Quantity 1 g
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-methyl-5-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.