Learn More
Accela Chembio Inc 2-chloro-4-methylphenylhydrazine Hydrochloride | 5g | 227958-97-6 | MFCD01861990 | 97+% | Shelf Life: 1440 Days | Light Sensitive/moisture Sensitive/+4
Supplier:  Accela Chembio Inc SY0135145G
Encompass_Preferred
2-chloro-4-methylphenylhydrazine Hydrochloride | 5g | 227958-97-6 | MFCD01861990 | 97+% | Shelf Life: 1440 Days | Light Sensitive/moisture Sensitive/+4
Catalog No. 50-251-909