Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-4-methylpyridine

Catalog No. 11632725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-632-725ML 25 mL
11-632-75ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-632-725ML Supplier Sigma Aldrich Supplier No. 11632725ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloro-4-picoline (2-Chloro-4-methylpyridine) has been used in the determination of gas-phase basicity of several relatively small organic molecules that are used in the MALDI.

Specifications

CAS 3678-62-4
Density 1.142 g/mL (at 25°C (literature))
Boiling Point 194°C to 195°C (lit.)
Molecular Formula C6H6ClN
Refractive Index n20/D 1.529 (literature)
Linear Formula C6H6ClN
MDL Number MFCD00023418
Quantity 25 mL
Synonym 2-Chloro-4-picoline
Molecular Weight (g/mol) 127.57
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.