Learn More
Accela Chembio Inc 2-chloro-4-methylpyrimidine | 25g | 13036-57-2 | MFCD00054434 | 97+% | Shelf Life: 1620 Days | Light Sensitive/moisture Sensitive/+4/n2
Catalog No. 50354300
Encompass_Preferred
2-chloro-4-methylpyrimidine | 25g | 13036-57-2 | MFCD00054434 | 97+% | Shelf Life: 1620 Days | Light Sensitive/moisture Sensitive/+4/n2
Catalog No. 50-354-300 Supplier Accela Chembio Inc Supplier No. SY00539325G
2-chloro-4-methylpyrimidine | 25g | 13036-57-2 | MFCD00054434 | 97+% | Shelf Life: 1620 Days | Light Sensitive/moisture Sensitive/+4/n2