Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-(methylsulfonyl)benzoic acid, 95%, Thermo Scientific™

Catalog No. JFD01500EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JFD01500EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JFD01500EA Supplier Thermo Scientific Chemicals Supplier No. JFD01500EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53250-83-2
Molecular Formula C8H6ClO4S
Molecular Weight (g/mol) 233.64
MDL Number MFCD00216496
InChI Key CTTWSFIIFMWHLQ-UHFFFAOYSA-M
Synonym 2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa
PubChem CID 735863
IUPAC Name 2-chloro-4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=C(C([O-])=O)C(Cl)=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 234.66
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-(methylsulfonyl)benzoic acid
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.