Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-methylsulphonylbenzoic acid, 95%, Thermo Scientific™

Catalog No. AC425010025
Click to view available options
Quantity:
1 g
2.5 g
Packaging:
Glass bottle

Catalog No. AC425010025

Supplier: Thermo Scientific Chemicals 425010025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 53250-83-2
Molecular Formula C8H6ClO4S
Molecular Weight (g/mol) 233.64
MDL Number MFCD00216496
InChI Key CTTWSFIIFMWHLQ-UHFFFAOYSA-M
Synonym 2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa
PubChem CID 735863
IUPAC Name 2-chloro-4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=C(C([O-])=O)C(Cl)=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 192.0°C to 193.0°C
Color Pink or White to Yellow
Loss on Drying 5% max.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 2.5 g
Solubility Information Solubility in water: insoluble
Formula Weight 234.66
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Chloro-4-methylsulphonylbenzoic acid, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.