Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-nitrobenzonitrile, 97%, Thermo Scientific™

Catalog No. XAX00005EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
XAX00005EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XAX00005EA Supplier Thermo Scientific Chemicals Supplier No. XAX00005EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28163-00-0
Molecular Formula C7H3ClN2O2
Molecular Weight (g/mol) 182.56
MDL Number MFCD00052203
InChI Key ZIGQFRQZYVQQDR-UHFFFAOYSA-N
Synonym 2-chloro-4-nitro-benzonitrile, benzonitrile, 2-chloro-4-nitro, 2-chloro-4-nitrobenzenecarbonitrile, pubchem18759, acmc-1ceg9, 2-chloro-4-nitrobenzonitril, chloronitrobenzenecarbonitrile, 3-chloro-4-cyanonitrobenzene, ksc497k1r
PubChem CID 141401
IUPAC Name 2-chloro-4-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=C(C#N)C(Cl)=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 182.57
Percent Purity 97%
Chemical Name or Material 2-chloro-4-nitrobenzonitrile
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.