Learn More
Medchemexpress LLC 2-chloro-4-nitrophenyl β-D-glucopyranoside | 120221-14-9 | MFCD00037475 | 98.0% | 335.69 g/mol | C12H14ClNO8 | 5 G
SDP
Catalog No. 5000301218
Encompass_Preferred
2-Chloro-4-nitrophenyl β-D-glucopyranoside is a chromogenic substrate for β-glucosidase that releases 2-chloro-4-nitrophenol upon enzymatic cleavage, producing a measurable color change for spectrophotometric assays and microbial growth media applications. It is supplied as a white to off-white solid with high purity suitable for enzymatic activity measurements and plate-based screening.
  • Releases chromophore upon enzymatic hydrolysis for colorimetric detection.
  • High purity for reliable enzymatic activity measurements.
  • Suitable for spectrophotometric assays and plate-based screening.
  • White to off-white solid, easy to weigh and dissolve in appropriate solvents.
  • Applicable in bacterial growth media to indicate β-glucosidase activity.
  • Molecular weight 335.69 g/mol and formula C12H14ClNO8.
Catalog No. 50-003-01218 Supplier Medchemexpress LLC Supplier No. HYW0398595G
2-Chloro-4-nitrophenyl β-D-glucopyranoside is a chromogenic substrate for β-glucosidase that releases 2-chloro-4-nitrophenol upon enzymatic cleavage, producing a measurable color change for spectrophotometric assays and microbial growth media applications. It is supplied as a white to off-white solid with high purity suitable for enzymatic activity measurements and plate-based screening.
  • Releases chromophore upon enzymatic hydrolysis for colorimetric detection.
  • High purity for reliable enzymatic activity measurements.
  • Suitable for spectrophotometric assays and plate-based screening.
  • White to off-white solid, easy to weigh and dissolve in appropriate solvents.
  • Applicable in bacterial growth media to indicate β-glucosidase activity.
  • Molecular weight 335.69 g/mol and formula C12H14ClNO8.